Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKCDBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584239 Supplier Invitrogen™ Supplier No. PA584239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHAVTVVTLL EKLASMLETL RERQGGLARR QGGLAGSVRR IQSGLGALSR SHDTTSNTLA QLLAKAERVS SHANAAQE Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 94%.

JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the 'Signalosome', which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKCDBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAVIN3
Gene Accession No. Q969G5
Gene Alias 3110015B12Rik; 6330514M23Rik; Caveolae-associated protein 3; cavin 3; CAVIN3; cavin-3; D3T-inducible gene 2 protein; Dig2; DIG-2; Dithiolethione-inducible gene 2 protein; dithiolethione-inducible gene-2; hSRBC; MGC20400; PKC-delta binding protein; Prkcdbp; protein kinase C delta binding protein; protein kinase C delta-binding protein; protein kinase C, delta binding protein; sdr-elated gene product that binds to c-kinase; sdr-related gene product that binds to c-kinase; serum deprivation response factor-related gene product that binds to C-kinase; SRBC
Gene Symbols CAVIN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAVIN3. Recombinant protein control fragment (Product #RP-101671).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112464
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.