Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKCDBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116740 Supplier Invitrogen™ Supplier No. PA5116740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat heart Immunogen sequence: SNTLAQLLAK AERVSSHANA AQERAVRRAA QVQRLEANHG LLVARGKLHV LLFKEEGEVP ASAFQKAPEP LGPADQSELG PEQLEA.

JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the Signalosome, which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKCDBP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CAVIN3
Gene Accession No. Q9Z1H9
Gene Alias 3110015B12Rik; 6330514M23Rik; Caveolae-associated protein 3; cavin 3; CAVIN3; cavin-3; D3T-inducible gene 2 protein; Dig2; DIG-2; Dithiolethione-inducible gene 2 protein; dithiolethione-inducible gene-2; hSRBC; MGC20400; PKC-delta binding protein; Prkcdbp; protein kinase C delta binding protein; protein kinase C delta-binding protein; protein kinase C, delta binding protein; sdr-elated gene product that binds to c-kinase; sdr-related gene product that binds to c-kinase; serum deprivation response factor-related gene product that binds to C-kinase; SRBC
Gene Symbols CAVIN3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 75-160 of human PRKCDBP (NP_6594772).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85332
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.