Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCE Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA502395S

Encompass
Encompass_Preferred

Catalog No. 50-254-1575


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

PRKCE Monoclonal antibody specifically detects PRKCE in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

PRKCE
Monoclonal
0.95 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P53370
Mouse
Affinity chromatography
RUO
11162
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI1B7
Unconjugated
NUDT6
5830406C15Rik; MGC125656; MGC125657; nPKC-epsilon; PKC e; PKC[e]; Pkce; Pkcea; PKCepsilon; PRKCE; PRKCH; protein kinase C; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C, epsilon; protein kinase C, eta; R75156
Full length human recombinant protein of human NUDT6 produced in HEK293T cell.
30 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.