Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCE, Rabbit anti-Human, Clone: G13-X, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15894
Specifications
PRKCE | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
MGC125656/MGC125657/PKCE/nPKC-epsilon | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
G13-X | |
ELISA (1:10000-1:50000) Immunocytochemistry Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
protein kinase C, epsilon | |
PRKCE | |
A synthetic peptide corresponding to amino acids 134-144 at N-terminus of human PRKCE. | |
RUO | |
5581 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction