Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKCSH Monoclonal Antibody (A6H9)
GREENER_CHOICE

Catalog No. PIMA544983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544983 Supplier Invitrogen™ Supplier No. MA544983
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRKCSH Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKCSH
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A6H9
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PRKCSH
Gene Accession No. P14314
Gene Alias 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60
Gene Symbols PRKCSH
Host Species Mouse
Immunogen Recombinant protein within human Glucosidase 2 subunit beta aa 51-250.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.