Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRKCSH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560419
Description
Immunogen sequence: SLTNHHFYDE SKPFTCLDGS ATIPFDQVND DYCDCKDGSD EPGTAACPNG SFHCTNTGYK PLYIPSNRVN DGVCDCCDGT DEYNSGVICE NTC Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
Specifications
PRKCSH | |
Polyclonal | |
Unconjugated | |
PRKCSH | |
80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P14314 | |
PRKCSH | |
Recombinant protein corresponding to Human PRKCSH. Recombinant protein control fragment (Product #RP-102945). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction