Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKCSH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559795 Supplier Invitrogen™ Supplier No. PA559795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPAEEDKMPP YDEQTQAFID AAQEARNKFE EAERSLKDME ESIRNLEQEI SFDFGPNGEF AYLYSQCYEL TTNEYVYRLC PFKLVSQKPK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKCSH
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRKCSH
Gene Accession No. P14314
Gene Alias 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60
Gene Symbols PRKCSH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRKCSH. Recombinant protein control fragment (Product #RP-102918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.