Learn More
Invitrogen™ PRKCSH Recombinant Rabbit Monoclonal Antibody (4Z3K3)

Description
Positive test controls include: HepG2, A-549, SKOV3. The target is usually found in the following locations: Endoplasmic reticulum. Immunogen sequence: PFKLVSQKPK LGGSPTSLGT WGSWIGPDHD KFSAMKYEQG TGCWQGPNRS TTVRLLCGKE TMVTSTTEPS RCEYLMELMT PAACPEPPPE APTEDDHDEL.
Specifications
Specifications
| Antigen | PRKCSH |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4Z3K3 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRKCSH |
| Gene Accession No. | P14314 |
| Gene Alias | 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.