Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKD2, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89999265 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-265 Supplier Abnova Corporation Supplier No. PAB31649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen PRKD2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:40000) Immunohistochemistry (1:100-300) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide).
Gene protein kinase D2
Gene Alias HSPC187/PKD2
Gene Symbols PRKD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 810-890 of human PRKD2 (phospho S876).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25865
Test Specificity This antibody detects endogenous levels of PRKD2 protein only when phosphorylated at S876.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.