Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110981 Supplier Invitrogen™ Supplier No. PA5110981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANNSPPSAQK SVLPTAIPAV LPAASPCSSP KTGLSARLSN GSFSAPSLTN SRGSVHTVSF LLQIGLTRES VTIEAQELSL SAVKDLVCSI VYQKFPECGF FGM Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 94%.

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role. The protein encoded by this gene is one of the PKC family members. This kinase can be activated rapidly by the agonists of G protein-coupled receptors. It resides in both cytoplasm and nucleus, and its nuclear accumulation is found to be dramatically enhanced in response to its activation. This kinase can also be activated after B-cell antigen receptor (BCR) engagement, which requires intact phopholipase C gamma and the involvement of other PKC family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKD3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prkd3
Gene Accession No. O94806
Gene Alias 4930557O20Rik; 5730497N19Rik; EPK2; nPKC-D1; nPKC-mu; nPKC-nu; PKCM; PKC-MU; Pkcnu; PKC-NU; PKD; PKD1; PKD3; PRKCM; Prkcn; PRKD1; Prkd3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D3; protein kinase EPK2; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D3
Gene Symbols Prkd3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PRKD3. Recombinant protein control fragment (Product #RP-91415).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23683
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.