Learn More
Promega PRKG1 (PKG) Kinase Enzyme System, 10microg
Catalog No. PRV4248 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4248 10 μg
1 options
Catalog No. PRV4248 Supplier Promega Supplier No. V4248
Recombinant full-length human PRKG1 (PKG) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PRKG1 is a homodimer, with each monomer containing a regulatory cGMP-binding domain and a catalytic domain.
  • Easily Screen and Profile PRKG1 (PKG) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C