Learn More
Promega PRKG1 (PKG) Kinase Enzyme System, 1mg
Catalog No. PRV6841 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6841 1 mg
1 options
Catalog No. PRV6841 Supplier Promega Supplier No. V6841
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PRKG1 (PKG) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PRKG1 is a homodimer, with each monomer containing a regulatory cGMP-binding domain and a catalytic domain.
  • Easily Screen and Profile PRKG1 (PKG) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C