Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501736011 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6011 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6011 Supplier Proteintech Group Inc Supplier No. 551381AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody is specific to PRKG2.

cGKII is thought to play a key role in a diverse set of physiological pathway. cGKII may mediate intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Edentification of the pathway that mediates intestinal fluid secretion by E. coli STa has potential medical implications because STa causes traveler′s diarrhea and about 50% of infant mortality in developing countries. Transfection experiments in human cells disclose that cGKII phosphorylates SOX9 and attenuates SOX9 function by inhibiting its nuclear entry. Impaired differentiation of cultured KMI chondrocytes can be restored by silencing Sox9 by RNA interference. cGKII is postulated to be a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.

Specifications

Antigen PRKG2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PRKG2
Gene Accession No. Q13237, Q61410, Q64595
Gene Alias cGK 2, cGK2, cGKII, PRKG2, PRKGR2
Gene Symbols Prkg2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19092, 25523, 5593
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.