Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRKRIP1 Monoclonal Antibody (4D11-3F11)
GREENER_CHOICE

Catalog No. PIMA522590
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522590 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522590 Supplier Invitrogen™ Supplier No. MA522590
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MASPAASSVR PPRPKKEPQT LVIPKNAAEE QKLKLERLMK NPDKAVPIPE KMSEWAPRPP PEFVRDVMGS SAGAGSGEFH VYRHLRRREY QRQDYMDAMA EKQKLYAEFQ KRLEKNKIAA EEQTAKRRKK RQKLKEKKLL AKKMKLEQKK QEGPGQPKEQ GSSSSAEASG TEEEEEVPSF TMGR.

Required for pre-mRNA splicing as component of the spliceosome (PubMed:28502770, PubMed:30705154). Binds double-stranded RNA. Inhibits EIF2AK2 kinase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKRIP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4D11-3F11
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PRKRIP1
Gene Accession No. Q9H875
Gene Alias C114; KRAB box domain containing 3; KRBOX3; likely ortholog of mouse C114 dsRNA-binding protein; PRKR interacting protein 1 (IL11 inducible); PRKR-interacting protein 1; PRKRIP1; Src41; TK5
Gene Symbols PRKRIP1
Host Species Mouse
Immunogen PRKRIP1 (AAH14298, 1 a.a. ~ 184 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79706
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.