Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRKRIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562331
Description
Immunogen sequence: VMGSSAGAGS GEFHVYRHLR RREYQRQDYM DAMAEKQKLD AEFQKRLEKN KIAAEEQTAK RRKKRQKL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Required for pre-mRNA splicing as component of the spliceosome (PubMed:28502770, PubMed:30705154). Binds double-stranded RNA. Inhibits EIF2AK2 kinase activity. [UniProt].
Specifications
PRKRIP1 | |
Polyclonal | |
Unconjugated | |
PRKRIP1 | |
C114; KRAB box domain containing 3; KRBOX3; likely ortholog of mouse C114 dsRNA-binding protein; PRKR interacting protein 1 (IL11 inducible); PRKR-interacting protein 1; PRKRIP1; Src41; TK5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79706 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H875 | |
PRKRIP1 | |
Recombinant protein corresponding to Human PRKRIP1. Recombinant protein control fragment (Product #RP-101250). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction