Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKX Polyclonal Antibody, Invitrogen™

Catalog No. PIPA544588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544588 Supplier Thermo Scientific Supplier No. PA544588

Rabbit Polyclonal Antibody

Peptide sequence: MEAPGLAQAA AAESDSRKVA EETPDGAPAL CPSPEALSPE PPVYSLQDFD Sequence homology: Human: 100%; Rat: 75%

PRKX Polyclonal antibody specifically detects PRKX in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKX
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRKX
Gene Accession No. P51817
Gene Alias cAMP-dependent protein kinase catalytic subunit PRKX; Pkare; PKA-related protein kinase; PKX1; PrKX; Protein kinase PKX1; protein kinase X; protein kinase X-linked; protein kinase, X-linked; putative serine/threonine kinase; serine/threonine-protein kinase PRKX
Gene Symbols PRKX
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PRKX
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5613
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.