Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKX Polyclonal Antibody, Invitrogen™

Catalog No. PIPA582527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582527 Supplier Thermo Scientific Supplier No. PA582527

Rabbit Polyclonal Antibody

Immunogen sequence: LDFHVKDLIK KLLVVDRTRR LGNMKNGAND VKHHRWFRSV DWEAVPQRKL KPPIVPKIAG DGDTSNFETY PENDWD

PRKX is a serine/threonine kinase related to the catalytic subunit of the cAMP-dependent protein kinases. PRKX is developmentally regulated and may be involved in renal epithelial morphogenesis. This protein may also be involved in macrophage and granulocyte maturation. Abnormal recombination between this gene and a related pseudogene on chromosome Y is a frequent cause of sex reversal disorder in XX males and XY females. Pseudogenes of this gene are found on chromosomes X, 15 and Y.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKX
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRKX
Gene Accession No. P51817
Gene Alias cAMP-dependent protein kinase catalytic subunit PRKX; Pkare; PKA-related protein kinase; PKX1; PrKX; Protein kinase PKX1; protein kinase X; protein kinase X-linked; protein kinase, X-linked; putative serine/threonine kinase; serine/threonine-protein kinase PRKX
Gene Symbols PRKX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRKX
Purification Method Anitgen affinity chromatography, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.