Learn More
PRL Mouse anti-Human, Clone: OTI6B1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500719

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.Specifications
PRL | |
Monoclonal | |
Unconjugated | |
PRL | |
GHA1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5617 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI6B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P01236 | |
PRL | |
Full length human recombit protein of human PRL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.