Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517986
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517986 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517986 Supplier Invitrogen™ Supplier No. PA517986
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs), which contains a PTP domain and a characteristic C-terminal prenylation motif. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This tyrosine phosphatase is a nuclear protein, but may primarily associate with plasma membrane. The surface membrane association of this protein depends on its C-terminal prenylation. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which implicated its role in the tumorigenesis. Studies in rat suggested that this gene may be an immediate-early gene in mitogen-stimulated cells. PRL phosphatases increase cell proliferation by stimulating progression from G1 into S phase. PRL-1 function is regulated in a cell cycle-dependent manner and implicate PRL-1 in regulating progression through mitosis, possibly by modulating spindle dynamics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PTP4A1
Gene Accession No. Q93096
Gene Alias AA415290; AU019864; C130021B01; DKFZp779M0721; HH72; PRL1; Prl-1; protein tyrosine phosphatase 4a1; protein tyrosine phosphatase type IVA 1; protein tyrosine phosphatase type IVA protein 1; protein tyrosine phosphatase type IVA, member 1; protein-tyrosine phosphatase 4a1; Protein-tyrosine phosphatase of regenerating liver 1; PTP(CAAX1); PTP(CAAXI); PTP4A1; PTP4A1 protein; PTP4A2; PTPCAAX1; RP5-1148A21.2
Gene Symbols PTP4A1
Host Species Goat
Immunogen Synthetic peptide sequence (KDSNGHRNNCCIQ) corresponding to the C-terminus amino acids of PTP4A1 (aa 161-173).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7803
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.