Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
| Antigen | PRL3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119377
![]() |
Invitrogen™
PA5119377 |
200 μL |
Each for $511.50
|
|
|||||
|
PIPA5121669
|
Invitrogen™
PA5121669 |
20 μL |
Each for $209.50
|
|
|||||
Description
PRL3 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs). PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This class of PTPs contain a PTP domain and a characteristic C-terminal prenylation motif. Studies of this class of PTPs in mice demonstrated that they were prenylated proteins in vivo, which suggested their association with cell plasma membrane. Overexpression of this gene in mammalian cells was reported to inhibit angiotensin-II induced cell calcium mobilization and promote cell growth. Two alternatively spliced variants exist. PRL-3 expression was found to be up-regulated in human liver carcinoma compared with normal liver. Its expression in colorectal cancers may contribute to the establishment of liver metastasis. In a different study, PRL-3 is found to be expressed in tumor metastasis and vasculature, regardless of the tumor source.Specifications
| PRL3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| Ptp4a3 | |
| AV088979; HPRL-3; potentially prenylated protein tyrosine phosphatase; pPtp4a3; PRL3; Prl-3; PRL-R; protein tyrosine phosphatase 4a3; protein tyrosine phosphatase type IVA 3; protein tyrosine phosphatase type IVA, member 3; protein-tyrosine phosphatase 4a3; Protein-tyrosine phosphatase of regenerating liver 3; Ptp4a3 | |
| Ptp4a3 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O75365 | |
| 11156 | |
| Recombinant protein Tyrosine Phosphatase Type IVA 3. The antigen corresponds to amino acid range 1-170 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title