Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549676
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549676 Supplier Invitrogen™ Supplier No. PA549676
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects transfected PRL3 protein.

The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs). PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This class of PTPs contain a PTP domain and a characteristic C-terminal prenylation motif. Studies of this class of PTPs in mice demonstrated that they were prenylated proteins in vivo, which suggested their association with cell plasma membrane. Overexpression of this gene in mammalian cells was reported to inhibit angiotensin-II induced cell calcium mobilization and promote cell growth. Two alternatively spliced variants exist. PRL-3 expression was found to be up-regulated in human liver carcinoma compared with normal liver. Its expression in colorectal cancers may contribute to the establishment of liver metastasis. In a different study, PRL-3 is found to be expressed in tumor metastasis and vasculature, regardless of the tumor source.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ptp4a3
Gene Accession No. O75365, Q9D658
Gene Alias AV088979; HPRL-3; potentially prenylated protein tyrosine phosphatase; pPtp4a3; PRL3; Prl-3; PRL-R; protein tyrosine phosphatase 4a3; protein tyrosine phosphatase type IVA 3; protein tyrosine phosphatase type IVA, member 3; protein-tyrosine phosphatase 4a3; Protein-tyrosine phosphatase of regenerating liver 3; Ptp4a3
Gene Symbols Ptp4a3
Host Species Rabbit
Immunogen Peptide sequence around aa. 161-166 (K-D-P-H-T) derived from Human PRL-3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11156, 19245, 362930
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.