Learn More
Invitrogen™ PRL3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total PTP4A3.
Specifications
Specifications
| Antigen | PRL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ptp4a3 |
| Gene Accession No. | O75365, Q9D658 |
| Gene Alias | AV088979; HPRL-3; potentially prenylated protein tyrosine phosphatase; pPtp4a3; PRL3; Prl-3; PRL-R; protein tyrosine phosphatase 4a3; protein tyrosine phosphatase type IVA 3; protein tyrosine phosphatase type IVA, member 3; protein-tyrosine phosphatase 4a3; Protein-tyrosine phosphatase of regenerating liver 3; Ptp4a3 |
| Gene Symbols | Ptp4a3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.