Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prlz/TPD52 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TPD52101AP

Description
Prlz/TPD52 Polyclonal antibody specifically detects Prlz/TPD52 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Prlz/TPD52 | |
Polyclonal | |
Unconjugated | |
Tpd52 | |
28 kDa calcium-dependent phosphoprotein; AI043038; calcium sensitive phosphoprotein (CSPP28); CSPP28; D52; hD52; LOC100058422; mD52; N8L; PC-1; pp28; PrLZ; prostate and colon associated protein; prostate leucine zipper; protein N8; si:ch211-203l9.2; si:ch211-203l9.3; TPD52; tumor promoter D52 protein; tumor protein D52; Unknown (protein for MGC:127812) | |
Synthetic peptide taken within amino acid region 1-50 on human Prostate leucine zipper isoform 5 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P55327, Q62393 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21985, 294900, 7163 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction