Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130275
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130275 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130275 Supplier Invitrogen™ Supplier No. PA130275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-30275 detects PRMT1 from human samples.

PRMT1 is a member of the protein arginine N-methyltransferase (PRMT) family. Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Increased expression of PRMT1 may play a role in various types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene PRMT1
Gene Accession No. Q99873
Gene Alias 6720434D09Rik; ANM1; arginine N-methyltransferase 1; AW214366; HCP1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HMT1 hnRNP methyltransferase-like 2; HMT1 hnRNP methyltransferase-like 2 isoform 3; HMT2; Hrmt1l2; interferon receptor 1-bound protein 4; IR1B4; Mrmt1; PRMT1; protein arginine methyltransferase 1; protein arginine N-methyltransferase 1
Gene Symbols PRMT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 316-347 amino acids from the C-terminal region of human PRMT1.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.