Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581570 Supplier Thermo Scientific Supplier No. PA581570
Add to Cart
Add to Cart

PRMT1 Polyclonal Antibody

PRMT1 Polyclonal Antibody for ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PRMT1
Gene Accession No. Q99873
Gene Alias 6720434D09Rik; ANM1; arginine N-methyltransferase 1; AW214366; HCP1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HMT1 hnRNP methyltransferase-like 2; HMT1 hnRNP methyltransferase-like 2 isoform 3; HMT2; Hrmt1l2; interferon receptor 1-bound protein 4; IR1B4; Mrmt1; PRMT1; protein arginine methyltransferase 1; protein arginine N-methyltransferase 1
Gene Symbols PRMT1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human PRMT1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.