Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT1 Rabbit anti-Human, Mouse, Rat, Clone: 10W10U4, Novus Biologicals™
SDP

Catalog No. NB167937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167937 100 μg
NB167938 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167937 Supplier Novus Biologicals Supplier No. NBP316423100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PRMT1 Monoclonal antibody specifically detects PRMT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PRMT1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10W10U4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PRMT1 (Q99873).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3276
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.