Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 112791AP150UL

Description
The PRMT1 gene encodes a protein arginine methyltransferase that functions as a histone methyltransferase specific for H4.Specifications
PRMT1 | |
Polyclonal | |
Unconjugated | |
PRMT1 | |
ANM1, HCP1, HMT2, HRMT1L2, IR1B4, PRMT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15469, 3276, 60421 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q63009, Q99873, Q9JIF0 | |
PRMT1 | |
PRMT1 Fusion Protein Ag1790 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction