Learn More
PRMT10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304190AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins, such as DTL/CDT2, BCL6 and PRDM1/BLIMP1. The SCF(FBXO11) complex mediates ubiquitination and degradation of BCL6, thereby playing a role in the germinal center B-cells terminal differentiation toward memory B-cells and plasma cells. The SCF(FBXO11) complex also mediates ubiquitination and degradation of DTL, an important step for the regulation of TGF-beta signaling, cell migration and the timing of the cell-cycle progression and exit. Binds to and neddylates phosphorylated p53/TP53, inhibiting its transcriptional activity. SCF(FBXO11) does not seem to direct ubiquitination of p53/TP53.Specifications
PRMT10 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6P2P2 | |
PRMT9 | |
Between 795 and 845 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRMT9 | |
Putative protein arginine N-methyltransferase 9, Putative protein arginine N-methyltransferase 10, 211-, PRMT9, PRMT10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
291947, 90826 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.