Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT2 Monoclonal Antibody (OTI5F9), TrueMAB™, OriGene
SDP

Catalog No. 501674446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4446 100 μL
50-254-0819 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4446 Supplier OriGene Supplier No. TA503587
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRMT is a member of the mammalian PRMT (protein arginine N-methyltransferase) family. The family consists of 7 members that share two large domains of homology. Outside of these domains, epitopes are identified and antibodies against all 7 PRMT members have been developed.These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins.

Specifications

Antigen PRMT2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI5F9
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRMT2
Gene Accession No. P55345
Gene Alias HRMT1L1
Gene Symbols PRMT2
Host Species Mouse
Immunogen Full length human recombit protein of human PRMT2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3275
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.