Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541711 Supplier Invitrogen™ Supplier No. PA541711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ERAGCCGYIP ANHVGKHVDE YDPEDTWQDE EYFGSYGTLK LHLEMLADQP Sequence homology: Human: 100%; Rat: 79%.

Arginine methyltransferase that methylates the guanidino nitrogens of arginyl residues in proteins such as STAT3, FBL, histone H4. Acts as a coactivator (with NCOA2) of the androgen receptor (AR)-mediated transactivation. Acts as a coactivator (with estrogen) of estrogen receptor (ER)-mediated transactivation. Enhances PGR, PPARG, RARA-mediated transactivation. May inhibit NF-kappa-B transcription and promote apoptosis. Represses E2F1 transcriptional activity (in a RB1-dependent manner). May be involved in growth regulation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT2
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRMT2
Gene Accession No. P55345
Gene Alias histone-arginine N-methyltransferase PRMT2; HMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 1; HMT1 hnRNP methyltransferase-like 1; Hrmt1l1; MGC111373; Prmt2; PRMT2 alpha; PRMT2 beta; PRMT2 gamma; protein arginine methyltransferase 2; protein arginine N-methyltransferase 2
Gene Symbols PRMT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PRMT2 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3275
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.