Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT3 Antibody (RAB-C372), FITC - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320143F
Description
PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for ELISASpecifications
PRMT3 | |
Monoclonal | |
FITC | |
EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
0.1 mL | |
Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
10196 | |
Store at 4°C in the dark. | |
IgG κ |
ELISA | |
RAB-C372 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction