Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT3 Monoclonal Antibody (PRMT3-367), Invitrogen™

Catalog No. PIMA125469
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125469 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125469 Supplier Thermo Scientific Supplier No. MA125469

Mouse Monoclonal Antibody

MA1-25469 is expected to cross react with mouse (100% conserved), rat (100% conserved), and human (92% conserved) due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT3
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone PRMT3-367
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation 0.01M PBS with 15mM sodium azide; pH 7.4
Gene Prmt3
Gene Accession No. O60678, O70467, Q922H1
Gene Alias 2010005E20Rik; 2410018A17Rik; AL033309; heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; PRMT3; protein arginine methyltransferase 3; protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3
Gene Symbols Prmt3
Host Species Mouse
Immunogen Synthetic peptide: TVHKNKKDPRSLIV conjugated to KLH, corresponding to amino acids 501-514 of Rat PRMT3
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10196, 71974, 89820
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.