Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRMT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582316
Description
Immunogen sequence: TFSHCKSEHQ FNIDSMVHKH GLEFYGYIKL INFIRLKNPT VEYMNSIYNP VPWEKEEYLK PVLEDDLLLQ FDVEDLYEPV SVPFSYPNGL SENTSVVEKL KHMEARALSA EAALARARED LQKMKQFAQD FVMHTDVRTC SSS.
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
Specifications
PRMT3 | |
Polyclonal | |
Unconjugated | |
Prmt3 | |
2010005E20Rik; 2410018A17Rik; AL033309; heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; PRMT3; protein arginine methyltransferase 3; protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60678 | |
Prmt3 | |
Recombinant protein corresponding to Human PRMT3. Recombinant protein control fragment (Product #RP-88804). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction