Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585433 Supplier Invitrogen™ Supplier No. PA585433
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (92%), Bovine (91%). Positive Control: IMR32, U87-MG, MCF-7, mouse brain, PRMT3-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Prmt3
Gene Accession No. O60678, Q922H1
Gene Alias 2010005E20Rik; 2410018A17Rik; AL033309; heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; PRMT3; protein arginine methyltransferase 3; protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3
Gene Symbols Prmt3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRMT3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10196, 71974
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.