Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRMT3 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142907
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_005779.1; NP_001138638.1; NP_001138639.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
Specifications
PRMT3 | |
Polyclonal | |
Unconjugated | |
Prmt3 | |
2010005E20Rik; 2410018A17Rik; AL033309; heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; PRMT3; protein arginine methyltransferase 3; protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
10196 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O60678 | |
Prmt3 | |
Peptide with sequence DPKTLISEPCGIKH (aa 408-421). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction