Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT4 Polyclonal Antibody
PRMT4 Polyclonal Antibody
Supplier: Thermo Scientific PA586984
Description
PRMT4 Polyclonal Antibody for Western Blot

Specifications
PRMT4 | |
Polyclonal | |
Unconjugated | |
Carm1 | |
Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10498, 363026, 59035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4AE70, Q86X55, Q9WVG6 | |
Carm1 | |
Recombinant Protein of human CARM1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction