Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596100 Supplier Invitrogen™ Supplier No. PA596100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSHYTSPSEN MWNTGSTYNL SSGMAVAGMP TAYDLSSVIA SGSSVGHNNL IPLANTGIVN HTHSRMGSIM STGIVQGSSG AQGSGGGSTS AHYAVNSQFT MGGPAISMAS PMSIPTNT Positive Samples: 22Rv1, A-431, Mouse spleen, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in DNA packaging, transcription regulation, signal transduction, mRNA stability, metabolism of nascent pre-RNA, and transcriptional activation. Tissue specificity: Overexpressed in prostate adenocarcinomas and high-grade prostatic intraepithelial neoplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Carm1
Gene Accession No. Q4AE70, Q86X55, Q9WVG6
Gene Alias Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4
Gene Symbols Carm1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 486-603 of human PRMT4/CARM1 (NP_9545921).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10498, 363026, 59035
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.