Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056641
Description
PRMT5 Polyclonal specifically detects PRMT5 in Human, Mouse samples. It is validated for Western Blot.Specifications
PRMT5 | |
Polyclonal | |
Western Blot 1:500-1:1000 | |
O14744 | |
PRMT5 | |
This antibody was developed against a mixture of synthetic peptides corresponding to human PRMT5 amino acid sequence. | |
0.2 mL | |
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm | |
10419 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS containing 0.1 mg/ml BSA. with 0.02% Sodium Azide | |
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction