Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT5 Antibody (OTI2H3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273604B
Description
PRMT5 Monoclonal specifically detects PRMT5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
PRMT5 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PRMT5 | |
Recombinant protein fragment corresponding to amino acids 238-499 of human PRMT5 (NP_006100) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunohistochemistry | |
OTI2H3 | |
Immunohistochemistry | |
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm | |
10419.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction