Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT5 Antibody (RAB-C136), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320131MFV610
Description
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for Flow CytometrySpecifications
PRMT5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
RAB-C136 | |
Flow Cytometry | |
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions. | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm | |
10419 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction