Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT5 Monoclonal Antibody (GT3610)
GREENER_CHOICE

Catalog No. PIMA531465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531465 Supplier Invitrogen™ Supplier No. MA531465
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (85%), Japanese Medaka (86%), Xenopus laevis (91%), Pig (100%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (98%). Positive Control: Jurkat, Raji, K562, THP-1, HL-60, NCI-H929, rat kidney. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PRMT5 (protein arginine N-methyltransferase 5) is an arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. PRMT5 specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins Sm D1 and Sm D3; such methylation is required for the assembly and biogenesis of snRNP core particles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT5
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GT3610
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PRMT5
Gene Accession No. O14744
Gene Alias 72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs
Gene Symbols PRMT5
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human PRMT5. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10419, 364382
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.