Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT5 Polyclonal Antibody, Huabio

Catalog No. 502544426
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4426 Supplier HUABIO RESEARCH INC Supplier No. R12043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRMT5 Polyclonal antibody specifically detects PRMT5 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PRMT5
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PRMT5
Gene Accession No. O14744, Q8CIG8
Gene Alias 72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal human Prtm5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10419, 27374
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.