Learn More
PRMT6 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | PRMT6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 |
| Gene | PRMT6 |
| Gene Accession No. | Q96LA8 |
| Gene Alias | AW124876; BB233495; BOS_3165; FLJ10559; FLJ51477; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; Protein arginine N-methyltransferase 6 |
| Gene Symbols | PRMT6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.