Learn More
Invitrogen™ PRMT6 Recombinant Rabbit Monoclonal Antibody (1Z10L6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542871
Description
Positive test controls include: HeLa, MCF7, NIH/3T3, Jurkat, Rat testis. The target is usually found in the following locations: Nucleus. Immunogen sequence: MLEWRLGFWS QVKQHYGVDM SCLEGFATRC LMGHSEIVVQ GLSGEDVLAR PQRFAQLELS RAGLEQELEA GVGGRFRCSC YGSAPMHGFA IWFQVTFPGG E.
Protein arginine N-methyltransferase 6 (PRMT6) PRMT6 demonstrates type I PRMT activity, capable of forming both omega-N monomethylarginine and asymmetric dimethylarginine derivatives on the recombinant glycine- and arginine-rich substrates. cts as a transcriptional coactivator of a number of steroid hormone receptors including ESR1, ESR2, PGR and NR3C1. Promotes fasting-induced transcriptional activation of the gluconeogenic program through methylation of the CRTC2 transcription coactivator. May play a role in innate immunity against HIV-1 in case of infection by methylating and impairing the function of various HIV-1 proteins such as Tat, Rev and Nucleocapsid protein p7.
Specifications
PRMT6 | |
Recombinant Monoclonal | |
1.34 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NZB1, Q96LA8 | |
PRMT6 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human PRMT6 (Q96LA8). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
1Z10L6 | |
Unconjugated | |
PRMT6 | |
AW124876; BB233495; FLJ10559; FLJ51477; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; protein arginine N-methyltransferase 6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
295384, 55170, 99890 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.