Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675624
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-624 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-624 Supplier Invitrogen™ Supplier No. PA141283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HeLa or NIH-3T3 whole cell lysate, antigen standard for PRMT7 (transient overexpression lysate).

Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene PRMT7
Gene Accession No. Q922X9, Q9NVM4
Gene Alias [Myelin basic protein]-arginine N-methyltransferase PRMT7; 4933402B05Rik; BC006705; FLJ10640; Histone-arginine N-methyltransferase PRMT7; KIAA1933; myelin basic protein-arginine N-methyltransferase; PRMT7; PRMT-7; protein arginine methyltransferase 7; protein arginine N-methyltransferase 7; RGD1304869
Gene Symbols PRMT7
Host Species Rabbit
Immunogen Amino acids 346-360 of human PRMT7.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214572, 54496
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.