Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142910
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142910 Supplier Invitrogen™ Supplier No. PA5142910
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_061896.1; NP_001171753.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PRMT7
Gene Accession No. Q922X9, Q9NVM4
Gene Alias [Myelin basic protein]-arginine N-methyltransferase PRMT7; 4933402B05Rik; BC006705; FLJ10640; Histone-arginine N-methyltransferase PRMT7; KIAA1933; myelin basic protein-arginine N-methyltransferase; PRMT7; PRMT-7; protein arginine methyltransferase 7; protein arginine N-methyltransferase 7; RGD1304869
Gene Symbols PRMT7
Host Species Goat
Immunogen Peptide with sequence C-PRFGEINDQDRTDR.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214572, 54496
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.