Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT7 Polyclonal Antibody, Invitrogen™

Catalog No. PI720143
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720143 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720143 Supplier Thermo Scientific Supplier No. 720143

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Bovine, Rabbit and Goat.

PRMT7 Polyclonal antibody specifically detects PRMT7 in Human, Non-human Primate samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT7
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2
Gene PRMT7
Gene Accession No. Q9NVM4
Gene Alias [Myelin basic protein]-arginine N-methyltransferase PRMT7; 4933402B05Rik; BC006705; FLJ10640; Histone-arginine N-methyltransferase PRMT7; KIAA1933; myelin basic protein-arginine N-methyltransferase; PRMT7; PRMT-7; protein arginine methyltransferase 7; protein arginine N-methyltransferase 7; RGD1304869
Gene Symbols PRMT7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRMT7 (aa 301-566)
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54496
Target Species Human, Non-human Primate
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.