Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541710 Supplier Invitrogen™ Supplier No. PA541710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YLEDYLTVRR GEEIYGTISM KPNAKNVRDL DFTVDLDFKG QLCETSVSND Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

Arginine methylation is a widespread posttranslational modification mediated by arginine methyltransferases, such as PRMT8. Arginine methylation is involved in a number of cellular processes, including DNA repair, RNA transcription, signal transduction, protein compartmentalization, and possibly protein translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRMT8
Gene Accession No. Q9NR22
Gene Alias arginine methyltransferase 8; heterogeneous nuclear ribonucleoprotein methyltransferase-like 4; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; HRMT1L4; PRMT8; protein arginine methyltransferase 8; protein arginine N-methyltransferase 4; protein arginine N-methyltransferase 8; protein arginine N-methyltransferase 8; protein arginine methyltransferase 8
Gene Symbols PRMT8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PRMT8 (aa 341-390).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56341
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.