Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143984
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143984 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143984 Supplier Invitrogen™ Supplier No. PA5143984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, human U-87MG whole cell. IHC: human meningeoma tissue, human meningeoma tissue, rat brain tissue, rat brain tissue. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Arginine methylation is a widespread posttranslational modification mediated by arginine methyltransferases, such as PRMT8. Arginine methylation is involved in a number of cellular processes, including DNA repair, RNA transcription, signal transduction, protein compartmentalization, and possibly protein translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene PRMT8
Gene Accession No. Q9NR22
Gene Alias arginine methyltransferase 8; heterogeneous nuclear ribonucleoprotein methyltransferase-like 4; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; HRMT1L4; PRMT8; protein arginine methyltransferase 8; protein arginine N-methyltransferase 4; protein arginine N-methyltransferase 8; protein arginine N-methyltransferase 8; protein arginine methyltransferase 8
Gene Symbols PRMT8
Host Species Rabbit
Immunogen E.coli-derived human PRMT8 recombinant protein (Position: M3-E171).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56341, 688502
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.