Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRMT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557845 Supplier Invitrogen™ Supplier No. PA557845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTKSLDIEIP KHIPERVSLV VTETVDAGLF GEGIVESLIH AWEHLLLQPK TKGESANCEK YGKVIPASAV IFGMAVECAE IRRHHRVGIK DIAGIHLPT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins, such as DTL/CDT2, BCL6 and PRDM1/BLIMP1. The SCF(FBXO11) complex mediates ubiquitination and degradation of BCL6, thereby playing a role in the germinal center B-cells terminal differentiation toward memory B-cells and plasma cells. The SCF(FBXO11) complex also mediates ubiquitination and degradation of DTL, an important step for the regulation of TGF-beta signaling, cell migration and the timing of the cell-cycle progression and exit. Binds to and neddylates phosphorylated p53/TP53, inhibiting its transcriptional activity. SCF(FBXO11) does not seem to direct ubiquitination of p53/TP53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRMT9
Gene Accession No. Q6P2P2
Gene Alias AI931714; Prmt10; PRMT9; protein arginine methyltransferase 10 (putative); protein arginine methyltransferase 9; Protein arginine N-methyltransferase 10; Protein arginine N-methyltransferase 9; putative protein arginine N-methyltransferase 10; putative protein arginine N-methyltransferase 9; RGD1306157
Gene Symbols PRMT9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRMT9. Recombinant protein control fragment (Product #RP-96723).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.